Erythrosin B, red powder C.I. 45430:2

Leverandør: MP Biomedicals
Warning

Synonymer: C.I. 45430:2

0219045025 0219045005
ICNA0219045025EA 2400 NOK
ICNA0219045025 ICNA0219045005
Erythrosin B, red powder C.I. 45430:2
Erythrosin B

Erythrosin B is a red, anionic xanthene dye.


Erythrosin B has been used as a contrast or counterstain to hematoxylin,crystal violet and other nuclear stains, in both plant and animal tissue.


It has been used as a contrast or counterstain to hematoxylin,crystal violet and other nuclear stains, in both plant and animal tissue.


Soluble in methyl cellosolve (20 mg/ml); slightly soluble in ethanol (2 mg/ml); very slightly soluble in water (0,7 mg/ml). Soluble in 0,05% methanol to give clear orange solution, in ammonium hydroxide (1 mg/ml) giving clear solution. Also spirit soluble.

Formel: C₂₀H₈I₄O₅
MDL nummer: MFCD00005044
CAS-nummer: 15905-32-5
EINECS: 240-474-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR