Nikotinamid-adenin dinukleotid (oksidert form) ≥98%, white powder cell culture reagent

Leverandør: MP Biomedicals

02194714.1 02194714.5 0219471401 0219471410
ICNA02194714.1EA 460 NOK
ICNA02194714.1 ICNA02194714.5 ICNA0219471401 ICNA0219471410
Nikotinamid-adenin dinukleotid (oksidert form) ≥98%, white powder cell culture reagent
Nikotinamid-adenin dinukleotid (oksidert form)

Storage: -20°C, desiccate
This is an ultrapure NAD, chromatographically purified to remove trace inhibitors.
β-NAD, a pyridine nucleotide and biologically active form of nicotinic acid, is a coenzyme necessary for the catalytic reaction of certain enzymes. It occurs in living cells primarily in the oxidized state. Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols. NAD usually acts as a hydrogen acceptor, forming NADH which then serves as a hydrogen donor in the respiratory chain.
Many metabolites and enzymes of biological interest are present in tissues at low concentrations. With the use of β-NAD as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling, much greater sensitivity for detection of these components is achieved. The reduced form, β-NADH, is fluorescent whereas β-NAD is not. This difference in fluorescence provides a sensitive fluorescent measurement of the oxidized or reduced pyridine nucleotides at concentrations down to 10-7 M.
Electron acceptor. β-NAD is a carrier for hydride ion, forming b-NADH. Hydride ion is enzymatically removed from a substrate molecule by the action of dehydrogenases such as malic dehydrogenase and lactic dehydrogenase. Such enzymes catalyze the reversible transfer of a hydride ion from malate or lactate to b-NAD to form the reduced product, b-NADH. Unlike b-NAD which has no absorbance at 340 nm, b-NADH absorbs at 340 nm (EmM = 6.22). The increase in absorbance at 340 nm with the formation of b-NADH is the basis for measurement of activity of many enzymes.


  • Cell Culture Reagent Grade
  • Extinction Coefficient (E1%): EmM = 18.0 (260 nm, pH 7) (Lit.)
  • Presentation: White Powder
  • Soluble in water (1%).

Formel: C₂₁H₂₇N₇O₁₄P₂
Smeltepunkt: 160 °C (lit.)
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036253
CAS-nummer: 53-84-9

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) 2-4
I max (water) 259 ± 3 nm
OD250/OD260 0.8 ± 0.15
OD280/OD260 0.2 ± 0.15


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR