Tris-(hydroksymetyl)aminometan

Leverandør: MP Biomedicals
Warning

0219485594 0219485580 0219485583 0219485590 0219485591
ICNA0219485594EA 16500 NOK
ICNA0219485594 ICNA0219485580 ICNA0219485583 ICNA0219485590 ICNA0219485591
Tris-(hydroksymetyl)aminometan
Tris-(hydroksymetyl)aminometan

Tris has been useful as buffers in a wide variety of biological systems. Uses include pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications. Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes). Tris does not precipitate calcium salts and is of value in maintaining solubility of manganese salts. It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid. It is an auxiliary material in pharmaceutical science.
Store at Room Temperature (15-30 °C). Store dessicated.


  • Molecular Biology Reagent Grade
  • Vapor Pressure: Low at ambient temperatures.(lit.)
  • Presentation: White Crystalline Powder
  • pKa: 8.1 at 25 °C(Lit.)


pH: 10 - 11.5 (5% aq soln)

Formel: NH₂C(CH₂OH)₃
Kokepunkt: 219…220 °C (10 mmHg)) (lit.)
Smeltepunkt: 167…172 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004679
CAS-nummer: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
Purity ≥99.95%
FTIR Conforms to Standard
pH (5% aq soln) 10-11.5
OD 260 nm (5% aq soln) ≤0.1
Loss on Drying ≤1%
Solubility (40% aq soln) Clear, Colorless Solution

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR