n-Heksadekan ≥99%, liquid

Leverandør: MP Biomedicals

0219521880 0219521890 0219521891
ICNA0219521880EA 900 NOK
ICNA0219521880 ICNA0219521890 ICNA0219521891
n-Heksadekan ≥99%, liquid
n-Heksadekan

Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.


Soluble in ether; insoluble in water.


Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)

Formel: H₃C(CH₂)₁₄CH₃
MDL nummer: MFCD00008998
CAS-nummer: 544-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR