Polyethyleneimine, M.W. 50,000 - 100,000 50% in aqueous solution, viscous solution

Leverandør: MP Biomedicals
Warning

Synonymer: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

0219544450 0219544480 0219544490 0219544491
ICNA0219544450EA 490 NOK
ICNA0219544450 ICNA0219544480 ICNA0219544490 ICNA0219544491
Polyethyleneimine, M.W. 50,000 - 100,000 50% in aqueous solution, viscous solution
Polyethyleneimine

Polyethylenimine is a branched chain polymer having a ratio of 1:2:1 of primary:secondary:tertiary amines with a branching site every 3 to 3,5 nitrogen atoms and a base structure of (CH₂CH₂NH)x. The nitrogen atoms must be protonated to achieve cationic charge, so the cationic charge density is pH dependent.


Polyethyleneimine is used for protein and nucleic acid precipitation, transfection assays, and to pretreat filters to increase the binding of proteins. Polyethyleneimine can also complex heavy metal ions. An effective permeabiliser of gram negative bacteria. Crosslinked polyethyleneimine is used as an enzyme carrier. Polyethyleneimine cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on Polyethyleneimine cellulose-coated plates.


Water soluble polymer used as a membrane lubricant; useful in filtering frozen cell membranes.


NB: Research Use Only (RUO).

Formel: (CH2CH2NH)n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00084427
CAS-nummer: 9002-98-6

Bestill nå

Spesifikasjon Testresultater

Identity Test Passes
FTIR Conforms to Standard
Concentration ~50% w/v

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR