Vitamin Diet Fortification Powder, MP Biomedicals
Leverandør: MP Biomedicals
0290465401
ICNA0290465401EA 3800 NOK
ICNA0290465401
Vitamin Diet Fortification Powder, MP Biomedicals
Rekvisita for dyr Dyreføde
Vitamin diet fortification mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise control of vitamin levels.
The ingredients of the vitamin diet fortification mixture are triturated in dextrose (1 kg is sufficient for 100 lbs of diet).