Guanidiniumtiocyanat ≥98%

Leverandør: MP Biomedicals
Danger

Synonymer: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

04820991
ICNA04820991EA 3700 NOK
ICNA04820991
Guanidiniumtiocyanat ≥98%
Guanidiniumtiocyanat

Research Grade Strong protein denaturant which inactivates nucleases approximately 2.5 times faster than guanidine hydrochloride.


pH: 5 -7 (6 M aq soln)

Formel: CH₅N₃·CHNS
MW: 118.16 g/mol
Smeltepunkt: 118
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013027
CAS-nummer: 593-84-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR