Flavobacterium meningosepticum Proline specific endopeptidase, MP Biomedicals

Leverandør: MP Biomedicals

08320821
ICNA08320821EA 14500 NOK
ICNA08320821
Flavobacterium meningosepticum Proline specific endopeptidase, MP Biomedicals
Enzymer

Proline specific endopeptidase specifically cleaves peptide bonds on the carboxy side of proline residues.


  • Isoelectric point (pI): 9,1
  • Contaminants: aminopeptidase, trypsin
  • Inhibitors: DFP, Z-Gly-Pro-CH₂Cl₂


It is useful for the determination of amino acid sequences of peptides and proteins containing proline residues. It is useful in cancer, neurology and diabetes research. It may be used as a drug target for neuropsychiatric diseases such as stress disorder, depression, and schizophrenia.


Unit definition: One unit causes the formation of one μmole of p-nitroaniline per minute under specific pH and temperature conditions.
Enzyme Activity: One unit is the enzyme activity which produces 1 μmol p-nitroaniline per minute at 30 °C, pH 7,0, from Z-Gly-Pro-pNA.

Bestill nå

SPESIFIKASJONER

Enzymnavn Proline specific endopeptidase
Environmentally Preferable
Kilde
Arter Flavobacterium meningosepticum
CAS-nr 72162-84-6
Molekylvekt ~78 kDa
Skjema Lyophilized powder
Lagringsforhold 2...8 °C
Enzymaktivitet 5,0 U/mg material
pH 9,1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR