Anti-C3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: MP Biomedicals
0855237
ICNA0855237EA 6275 NOK
ICNA0855237
Anti-C3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Peroxidase-Conjugated Goat IgG Fraction to Human Complement C3 is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to human complement C3 and buffer salts.

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR