Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: MP Biomedicals
0855526
ICNA0855526EA 4150 NOK
ICNA0855526
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
This product is fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat affinity purified F(ab')2 fragments to mouse immunoglobulins (IgG,IgA,IgM) and buffer salts.The total protein is measured by absorbance (A)280 nm); F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the affinity purified F(ab')2fragments is standardized with an in-house control by immunoelectrophoresis. The affinity purified F(ab')2 fragment is tested for purity at 10 mg/mL using immunoelectrophoresis. The product is goat F(ab')2; no trace of albumin, whole molecule IgG or Fc is detected.More Product Information