Human IgG Isotype Control

Leverandør: MP Biomedicals
0855836
ICNA0855836EA 1275 NOK
ICNA0855836
Human IgG Isotype Control
Antistoffer
This product is the lyophilized powder of human IgG positive control and buffer salts. IgG is a purified antigen. Immunoglobulin G (IgG) is one of the most abundant proteins in serum. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions: to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities inMore Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antistofftype Control
Konjugering Unconjugated
Reaktivitet
Vert Human
Isotype IgG
Skjema Lyophilized from PBS, no azide
Rensing Delipidation, ammonium sulfate fractionation and anion exchange chromatography. The resulting IgG fraction is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed and lyophilized.
Konsentrasjon Total Protein: 10.5 mg/mL; IgG Concentration: 10.0 mg/mL. Packaged as 10 mg/mL
Frakttemp. Cold Pack

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR