Human IgA Isotype Control

Leverandør: MP Biomedicals
0855906
ICNA0855906EA 7850 NOK
ICNA0855906
Human IgA Isotype Control
Antistoffer
Product is the lyophilized powder of human serum IgA and buffer salts. The product is tested for purity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is human serum IgA; no trace of IgG, IgM or non-immunoglobulin serum proteins is detected. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 5 mg of human serum IgA is obtained upon reconstitution.
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antistofftype Control
Konjugering Unconjugated
Reaktivitet
Vert Human
Isotype IgA
ELISA Yes
Skjema Lyophilized
Rensing Human IgA is purified from pooled human serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 μm filter, vialed and lyophilized.
Konsentrasjon Total Protein: 5.41 mg/mL; Volume: 1 mL
Frakttemp. RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR