MacConkey agar, MP Biomedicals
Leverandør: MP Biomedicals
091006017
ICNA091006017EA 1375 NOK
ICNA091006017
MacConkey agar, MP Biomedicals
Mikrobiologiske medier Media, dehydrert for mikrobiologi
A preparation for the isolation of enteric bacilli based on the fermentation of lactose.
Ready for CE IVD labeling of clinical applications.
NB: Research use only (RUO).