Cell culture media, Ham's F10 (10X), MP Biomedicals
Leverandør: MP Biomedicals
091440049
ICNA091440049EA 450 NOK
ICNA091440049
Cell culture media, Ham's F10 (10X), MP Biomedicals
Media, Cellekultur
Ham's nutrient mixtures were developed for the clonal growth of several chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells.
  • Ideal for mammalian cell culture
  • Available as clear, orange solution
  • pH of 5,5 to 6,1
  • To be stored at 4 °C
Ham's F-10 has demonstrated satisfactory growth of human diploid cells, white blood cells for chromosomal analysis and primary rat, rabbit and chicken tissue explants. Both F-10 and F-12 are formulated for use with or without serum, depending on the type of cells being cultured.