Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals

Leverandør: MP Biomedicals

091810454
ICNA091810454EA 315 NOK
ICNA091810454
Hanks' Balanced Salts (Modified), 1X, without Magnesium and Calcium, MP Biomedicals
Buffere Buffere, cellekultur

Used to minimise cell attachment or aggregation.


  • Sterile
  • Suitable for cell culture
  • Formulation: Inorganic Salts: Potassium Chloride [KCl] 400.00000 mg/L, Potassium Phosphate Monobasic [KH2PO4] 60.00000 mg/L, Sodium Bicarbonate [NaHCO3] 350.00000 mg/L, Sodium Chloride [NaCl] 8000.00000 mg/L, Sodium Phosphate Dibasic [Na2HPO4] 47.50000 mg/L, Dextrose 1000.00000 mg/L, Phenol Red Sodium Salt 17.00000 mg/L
  • pH: 6,9-7,4
  • Osmolality 258 - 298 mM/kg


Calcium and magnesium support cell adhesion. Hanks' Balanced Salts (Modified), without Magnesium and Calcium, is used to minimise cell attachment or aggregation.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR