Human Serum, Pooled, MP Biomedicals
Leverandør: MP Biomedicals
092930949
ICNA092930949EA 4625 NOK
ICNA092930949
Human Serum, Pooled, MP Biomedicals
Serum
Human pooled serum is used as a media component to neutralize cell free virus. It's used as a reagent in liposome binding assay and in preparation of sperm.
  • Obtained from normal clotted blood
  • Liquid media
  • Store at —20 °C
  • Media component for cell free virus, liposome binding assay, preparation of sperm
Each donor unit of plasma used in the preparation of this material was tested and found negative for HBsAg, HIV 1/2, anti-HCV, HCV-NAT, HIV-1 NAT, and syphilis by FDA regulation.