Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedicals
Leverandør: MP Biomedicals
092931949
ICNA092931949EA 3575 NOK
ICNA092931949
Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedicals
Serum
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
  • Collected from Type AB human blood
  • Liquid media
  • Store at —20 °C
It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.