Cleaning solution, ES 7X™, ready to use, MP Biomedicals

Leverandør: MP Biomedicals
097668193
ICNA097668193EA 655 NOK
ICNA097668193
Cleaning solution, ES 7X™, ready to use, MP Biomedicals
Rengjøringsløsninger Detergenter og overflatevasking
Ready to use 5% cleaning solution.

  • Soluble in water
  • Store at 15 to 30 °C

Leveringsinformasjon: Supplied with a hand-pump for convenient dispensing.
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR