Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Leverandør: MP Biomedicals
112350200
ICNA112350200EA 3450 NOK
ICNA112350200
Ethidium bromide Greenbag™ disposal kit, MP Biomedicals
Elektroforese reagenser Reagenser for fjerning av fargeløsning
The EtBr Greenbag™ Disposal Kit allows rapid and trouble-free concentration of ethdium bromide from large volumes of solutions into a small 'teabag' containing activated carbon. The Greenbag™ is then conveniently disposed of along with other solid hazardous wastes.
One kit has the capacity to remove 500 mg of ethidium bromide from solutions (10 mg EtBr/bag), enough to satisfy the requirements of most laboratories for a year.