Dehydrated culture media, 2xYT-broth, MP Biomedicals
Leverandør: MP Biomedicals
113012026 113012231 113012042 113012041 113012032 113012021 113012036 113012046
ICNA113012026EA 0 NOK
ICNA113012026 ICNA113012231 ICNA113012042 ICNA113012041 ICNA113012032 ICNA113012021 ICNA113012036 ICNA113012046
Dehydrated culture media, 2xYT-broth, MP Biomedicals
Mikrobiologiske medier Media, dehydrert for mikrobiologi
2xYT-broth is a nutritional rich medium providing nitrogen and growth factors optimised for the growth of recombinant Escherichia coli strains infected with the M13 bacteriophage or other filamentous single stranded DNA bacteriophages thus allowing the bacteriophage to reproduce in large quantities without exhausting the host cells.
  • Available as capsules or powder form
YT media formulations are optimised for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages.