Dehydrated culture media, DOB media, MP Biomedicals
Leverandør: MP Biomedicals
114025032 114025012 114025022
ICNA114025032EA 7225 NOK
ICNA114025032 ICNA114025012 ICNA114025022
Dehydrated culture media, DOB media, MP Biomedicals
Mikrobiologiske medier Media, dehydrert for mikrobiologi
DOB is a component of Synthetic Defined Media (SD), which is used for growth of S. cerevisiae.
  • A complete yeast nitrogen base with a carbon source
A complete supplemented synthetic defined medium is easily made by mixing DOB and CSM, a complete supplement mixture containing amino acids, adenine and uracil.
If you do not want to purchase a pre-made SD or SDA medium, or the combination of amino acids that you need is unavailable, you can easily create your own SD or SDA by combining DOB or DOBA with the appropriate amino acid mixture.
DOB is a complete yeast nitrogen base with a carbon source. A complete supplemented synthetic defined medium is easily made by mixing DOB and CSM, a complete supplement mixture containing amino acids, adenine and uracil.
Add 26,7 g/L DOB plus the amino acid mixture of you choice to make selective SD medium.
Applications includes: Yeast cell culture, gene expression, transformation, gene function; gene regulation.
Sertifiseringer: Molecular Biology Certified™.