Fast DNA™ SPIN kit for soil, MP Biomedicals
Leverandør: MP Biomedicals
116560200
ICNA116560200EA 6050 NOK
ICNA116560200
Fast DNA™ SPIN kit for soil, MP Biomedicals
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
The Fast DNA™ SPIN Kit for Soil is designed to efficiently isolate bacterial, fungal, plant and animal genomic DNA from soil and other environmental samples. It quickly and efficiently isolates PCR-ready genomic DNA directly from soil samples in less than 60 minutes.
  • Isolation of DNA from cells present in soil or other environmental samples
  • Rapid and reproducible sample lysis with the FastPrep®-24 or FastPrep® FP120 instrument
  • Easily isolate DNA from a variety of organisms in many different types of soil
  • Lyse and isolate DNA in less than 60 minutes
  • No hazardous organic reagents required
The FastDNA® SPIN Kit is designed for use with any FastPrep® instrument. Plant and animal tissues, bacteria, algae, fungi spores and other members of a soil population are easily lysed within 40 seconds. These benchtop devices use a unique, optimised motion to homogenise samples by simultaneous impaction with lysing matrix particles.
For research purposes only.
Leveringsinformasjon: Kit contains Lysing Matrix E, 50×2,0 ml tubes; Sodium Phosphate Buffer, 60 ml; MT Buffer, 8 ml; PPS Solution, 25 ml; Binding Matrix, 66 ml; SPIN Modules, 50 each; Catch Tubes, 50 each; Concentrated SEWS-M, 12 ml; DES, 20ml; and BBS Gel Loading Dye, 200 µl.