Lysing kits for Fastprep®-24 5G, Lysing Matrix C, MP Biomedicals
Leverandør: MP Biomedicals
116932050 116912500 116912100 116912050 116932005 116540432 116540433 116932025 116952050 116982010
ICNA116932050EA 6275 NOK
ICNA116932050 ICNA116912500 ICNA116912100 ICNA116912050 ICNA116932005 ICNA116540432 ICNA116540433 ICNA116932025 ICNA116952050 ICNA116982010
Lysing kits for Fastprep®-24 5G, Lysing Matrix C, MP Biomedicals
Homogenisatorer Homogenisatorer, bordmodell
Each impact-resistant 2 ml tube contains 1 mm silica spheres. Lysing Matrix C tubes have red caps and are found in the FastRNA™ Pro Red Kit and FastProtein Red Matrix. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for moulds, spores and filamentous algae.