Cleaning solution, DNA-OFF™, MP Biomedicals
Leverandør: MP Biomedicals
11QD0500
ICNA11QD0500EA 1125 NOK
ICNA11QD0500
Cleaning solution, DNA-OFF™, MP Biomedicals
Rengjøringsløsninger Dekontaminanter
DNA-OFF™ solution contains a surfactant plus an agent that destroys DNA intended for use at PCR workstations including countertops, apparatus and pipettors.
  • Used for eliminating DNA from all surfaces
  • It is stable and heat resistant
DNA-OFF™ is a ready to use non-corrosive solution and does not contain carcinogenic compounds.