Handheld digital microscopes, special light, stroboscopic
Leverandør: Dino-Lite / IDCP B.V.
AM3713TB AM3715TB
IDCPAM3713TBEA 5925 NOK
IDCPAM3713TB IDCPAM3715TB
Handheld digital microscopes, special light, stroboscopic
Mikroskop
Portable microscopes with stroboscopic technology to capture fast moving objects.
  • Perfect pictures thanks to reduced motion blur, even under higher magnification
  • Stroboscopic light mode can be switched on and off
  • High speed USB 2.0 output
  • Edge series model with high-quality optics provides a very sharp, bright and natural colour image with very low aberration and vignetting
  • AM3715TB features exchangeable caps and an external trigger to synchronise picture capturing
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10