Digital Microscope
Leverandør: Dino-Lite / IDCP B.V.
AM4117MT-G2FBW
IDCPAM4117MT-G2FBWEA 14800 NOK
IDCPAM4117MT-G2FBW
Digital Microscope
Mikroskop
With 465 nm excitation light and a 510 - 545 nm emission bandpass filter range, AM4117MT-G2FBW is the ideal solution for the selective isolation and observation of samples with green fluorescence.
  • Green fluorescence
  • High sensitivity sensor
  • Adjustable LED intensity
  • Robust metallic housing
  • Interchangeable caps
The interchangeable caps provide adaptability to numerous applications with alternative lighting or object interface. The interchangeable front caps are designed to be used with Dino-Lite Edge and EdgePLUS series models. The caps provideMore Product Information