Digital Microscope
Leverandør: Dino-Lite / IDCP B.V.
AM8117MZT
IDCPAM8117MZTEA 16825 NOK
IDCPAM8117MZT
Digital Microscope
Mikroskop
Taking image quality to the next level, the 8-megapixel, this digital microscope delivers high resolution and color fidelity for a natural-looking and detailed imaging experience.
  • High optical resolution
  • Enhanced flexible LED Control (eFLC)
  • Adjustable polariser
  • Robust metal housing
  • 10× ~ 220× magnification (Edge shading occurs below 25× when previewing in 16:9 windows.) french, spanish and more
Includes a polariser for reducing unwanted reflections as well as eFLC for maximising lighting adjustability.
2 years warranty.
Forpakning: N3C-C (closeMore Product Information