Digital Microscope
Leverandør: Dino-Lite / IDCP B.V.
AM8917MZT
IDCPAM8917MZTEA 21550 NOK
IDCPAM8917MZT
Digital Microscope
Mikroskop
Pushing the limits of handheld digital microscopy, the 8-megapixel AM8917MZT is a high-resolution digital microscope capable of revealing the minute details of the target. The microscope also delivers outstanding colour fidelity, enhancing the imaging experience with detailed and natural-looking results. Ready to perform a wide range of tasks, AM8917MZT offers a polarizer as well as DPQ, EDOF, EDR, AMR, and eFLC.
  • High optical resolution
  • Enhanced flexible LED control (eFLC)
  • Automatic magnification reading (AMR)
  • Robust metal housing
  • Extended depth of field (