Microscope stand, rolling, Dino-Lite MS-W1
Leverandør: Dino-Lite / IDCP B.V.
MS-W1
IDCPMS-W1EA 1550 NOK
IDCPMS-W1
Microscope stand, rolling, Dino-Lite MS-W1
Stativer
The MS-W1 stand with HD-M1 holster is a special stand for cylindrical surfaces or to roll on flat surfaces.
The arcs on the MS-W1 enable the user to easily examine cylindrical objects such as printing rolls. The barrel rollers at the bottom of the arcs are wrapped in rubber which helps to prevent the stand from scratching or harming the surfaces it is examining. The MS-W1 can be used to examine tubular objects of at least 7 centimeters in diameter. With the two barrel rollers, the MS-W1 stand can also be rolled forward or backward making it a great solution for surface inspection; such asMore Product Information