Digital Microscope
Leverandør: Dino-Lite / IDCP B.V.
WF7915MZT
IDCPWF7915MZTEA 20850 NOK
IDCPWF7915MZT
Digital Microscope
Mikroskop
With the use of the latest, cutting-edge optics, a brand new 5 megapixel sensor and several special features, it has a marvel of technology and the best choice for the high-demanding professional. It offers superb image quality and color reproduction in a robust, compact and appealing housing.
  • Wireless
  • Extended depth of field (EDOF)
  • Extended dynamic range (EDR)
  • Flexible LED control (FLC)
  • Automatic magnification reading (AMR)
With the Extended Dynamic Range (EDR) feature, the details of darker or brighter areas within the object can be revealed byMore Product Information