Evaporating flasks
Leverandør: IKA
1905700
IKAA1905700EA 2175 NOK
IKAA1905700
Evaporating flasks
Kolber Kolber, rotasjonsfordamper
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard IKA® rotary evaporators; with white labelling area.