Bad
Leverandør: IKA
20004382 20004383
IKAA20004382EA 6950 NOK
IKAA20004382 IKAA20004383
Bad
Bad Vannbad, uten sirkulasjon
NB: Note: The product is a rectangular plastic bath without a thermostat, the thermostatic controller must be ordered separately.