Laboratory reactor systems, LR-2.ST the versatile
Leverandør: IKA
20013293
IKAA20013293EA 0 NOK
IKAA20013293
Laboratory reactor systems, LR-2.ST the versatile
Laboratoriereaktor systemer
Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. The system is characterised by the agitator mounting, which allows a safe transfer of the higher motor torque.
  • Suitable for vacuum operation
  • Solvent- and temperature-resistant perfluoro elastomer (FFPM) seals meet the sample
  • Infinitely variable speed
  • Torque trend display for measuring changes in viscosity
  • Microprocessor-controlled speed regulation, enables steady speed, also under load
  • Removable WiCo (wireless
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Bruksvolum 500 - 2000 ml
Temperaturområde RT*...230 °C
Hastighetsområde 8 - 290 min⁻¹
Maks. viskositet 70000 mPas
Vakuum 25 mbar
Materiale i kontakt med medium Borosilicate glass, FFPM, PTFE, 1.4571 steel
B×D×H 460×430×1240 mm
Vekt 25 kg