Magnetic hotplate stirrer, IKA Plate RCT digital
Leverandør: IKA
25004601
IKAA25004601EA 15100 NOK
IKAA25004601
Magnetic hotplate stirrer, IKA Plate RCT digital
Omrørere Magnetrørere med varmeplate
The IKA Plate (RCT digital) magnetic hotplate stirrer has an aluminium alloy plate, suitable for optimum heat transfer. It features a glass housing surface for optimum cleaning and chemical resistance. The unit features a strong motor for a high range of speeds, integrated timer and counter function supports the monitoring of sensitive chemical reactions.
  • SmartTemp® hot surface warning to protect against burns
  • Connector for electronic temperature control (via optional PT1000 probe)
  • Regulation of the plate can be optimised by regular firmware updates