tempmate® B3 Temperature Data Logger
Leverandør: tempmate
100530
IMEC100530EA 6125 NOK
IMEC100530
tempmate® B3 Temperature Data Logger
Dataloggere
Ultra robust mini logger, the size of a button cell, for recording temperature measurements with up to five years recording duration.
  • Ultra compact
  • Ultra light weight (4 g)
  • Calibration certificates optionally available
  • Splash waterproof
  • Cost-efficient and reliable
tempmate.®-B series temperature data loggers are ideal whenever your goods will be transported and stored over a long period of time and you have to make sure, that the quality has not been compromised. Its ultra-compact dimensions, super light weight and the economical pricing make ourMore Product Information