tempmate® B4 Data Logger for Temperature and Humidity
Leverandør: tempmate
100540
IMEC100540EA 6250 NOK
IMEC100540
tempmate® B4 Data Logger for Temperature and Humidity
Dataloggere
Ultra robust mini logger, the size of a button cell, for recording temperature and humidity measurements with up to fiive years recording duration.
  • Ultra compact
  • Ultra light weight (4 g)
  • Two channel data logger
  • Calibration certificates optionally available
  • Splash waterproof
  • Cost-efficient and reliable
tempmate.®-B series temperature data loggers are ideal whenever your goods will be transported and stored over a long period of time and you have to make sure, that the quality has not been compromised. Its ultra-compact dimensions, super light weightMore Product Information