tempmate.®-M1 Multi-Use Temperature Data Logger
Leverandør: tempmate
200100
IMEC200100EA 460 NOK
IMEC200100
tempmate.®-M1 Multi-Use Temperature Data Logger
Dataloggere
Intuitive multi-use temperature logger for reliable low-cost cold chain monitoring. For temperature recording with internal or external temperature sensor, multifunction display and automatic PDF report generation.
  • Product-related temperature monitoring along the entire cold chain, in warehouses or fridges
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • Free available software for individual logger configuration
  • Detailed PDF report including graph and table
  • Multifunction display with battery indicator