tempmate.®-i1 Single Use Indicator, 2 to 8 °C Start Delay 10 mins, No Alarm Delay
Leverandør: tempmate
I1CS110
IMECI1CS110EA 4300 NOK
IMECI1CS110
tempmate.®-i1 Single Use Indicator, 2 to 8 °C Start Delay 10 mins, No Alarm Delay
Dataloggere
Single use temperature indicator for reliable low-cost cold chain monitoring.
  • Product-related temperature monitoring along the entire cold chain
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • No additional hardware or software required
  • Storage time 4 years
  • Intelligent LEDs for status monitoring
tempmate.®-i1 indicators are specially designed for temperature monitoring in pharmaceutical cold chain logistics. As preprogrammed device tempmate.®-i1 can be instantly used to control your temperature sensitiveMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Grensesnitt
Beskyttelsesklasse IP 67/NEMA 6
Vekt 10 g