Anti-Antithrombin III Goat Polyclonal Antibody
Leverandør: ImmunoReagents
IR1370
IMMRIR1370EA 1025 NOK
IMMRIR1370
Anti-Antithrombin III Goat Polyclonal Antibody
Antistoffer
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  
  • Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information