Anti-IgM µ Goat Polyclonal Antibody (Biotin (NHS-LC))

Leverandør: ImmunoReagents
IR1779
IMMRIR1779EA 1125 NOK
IMMRIR1779
Anti-IgM µ Goat Polyclonal Antibody (Biotin (NHS-LC))
Antistoffer
ImmunoReagents Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents antibodies are suitableMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IgM
Antigen-navn Immunoglobulin M mu-chain
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Biotin (NHS-LC)
Reaktivitet Human
Vert Goat
Isotype IgM
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Epitop µ-chain
Immunogen Human IgM, µ chain
Format Purified IgG
Skjema Clear, colorless liquid
Rensing Affinity purified using solid phase Human IgM
Konsentrasjon 1,50 mg/ml (E 1% at 280 nm = 13,0)
Kyssabsorpsjon No
Lagringsbuffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
Lagringstemp 2…8 °C
Frakttemp. 2…8 °C
Uttestet applikasjon Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR