Anti-IgG Goat Polyclonal Antibody
Leverandør: ImmunoReagents
IR1908
IMMRIR1908EA 1750 NOK
IMMRIR1908
Anti-IgG Goat Polyclonal Antibody
Antistoffer
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
  • Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagentsMore Product Information