Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))
Leverandør: ImmunoReagents
IR1921
IMMRIR1921EA 2200 NOK
IMMRIR1921
Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))
Antistoffer
ImmunoReagents Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.
  • Origin: US
ImmunoReagents antibodies are suitableMore Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol IgG
Environmentally Preferable
Antigen-navn Immunoglobulin G
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Biotin (NHS-LC)
Reaktivitet Mouse
Vert Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Immunogen Mouse IgG
Format Purified IgG
Skjema Clear, colorless liquid
Rensing Affinity purified using solid phaseMouse IgG
Konsentrasjon 1,50 mg/ml (E 1% at 280 nm = 13,0)
Kyssabsorpsjon Yes
Lagringsbuffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
Lagringstemp 2…8 °C
Frakttemp. 2…8 °C
Uttestet applikasjon Fluorescence-Activated Cell Sorting