Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))

Leverandør: ImmunoReagents
IR2147
IMMRIR2147EA 1950 NOK
IMMRIR2147
Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))
Antistoffer
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualised, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies areMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antigen-navn Immunoglobulin G
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Biotin (NHS-LC)
Reaktivitet Rabbit
Vert Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Antistoff fragment F(ab')2
Immunogen Rabbit IgG
Format Purified IgG
Skjema Clear, colourless liquid
Rensing Affinity purified using solid phaseRabbit IgG
Konsentrasjon 1,50 mg/ml (E 1% at 280 nm = 13,0)
Kyssabsorpsjon Yes
Lagringsbuffer 10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2, 1 % (w/v) BSA, protease/IgG free
Lagringstemp 2...8 °C
Frakttemp. 2...8 °C
Uttestet applikasjon Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR