Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))
Leverandør: ImmunoReagents
IR2259
IMMRIR2259EA 3250 NOK
IMMRIR2259
Anti-IgG Goat Polyclonal Antibody (Biotin (NHS-LC))
Antistoffer
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualised, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.
  • Origin: US
ImmunoReagents' antibodies areMore Product Information