Anti-IgG Rabbit Polyclonal Antibody

Leverandør: ImmunoReagents
IR2636
IMMRIR2636EA 1575 NOK
IMMRIR2636
Anti-IgG Rabbit Polyclonal Antibody
Antistoffer
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antigen-navn Immunoglobulin G
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Rat
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Immunogen Rat IgG
Format Purified IgG
Skjema Clear, colorless liquid, 0.2 µm filtered
Rensing Affinity purified using solid phaseRat IgG
Konsentrasjon > 4,5 mg/ml (E 1% at 280 nm = 13,0)
Kyssabsorpsjon No
Lagringsbuffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
Lagringstemp 2...8 °C
Frakttemp. 2-8 °C
Uttestet applikasjon Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR