Anti-IgG (H+L) F(ab)'2 Goat Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: ImmunoReagents
IR2937
IMMRIR2937EA 11825 NOK
IMMRIR2937
Anti-IgG (H+L) F(ab)'2 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
The ImmunoReagents' Allophycocyanin (APC) is an intensely bright phycobiliprotein isolated from red algae that exhibits far-red fluorescence with high quantum yields. It is excited by laser lines at 594 and 633 nm, with an absorbance maximum at 650 nm and a fluorescence emission peak at 660 nm. APC is a large molecule that is chemically crosslinked to use in fluorescence-based detection, primarily for flow cytometry, microarray assays, ELISAs, and other applications that require high sensitivity but not photostability.
This conjugate is suitable for immunomicroscopy and flow cytometry.More Product Information