Blender bags, BagLight®
Leverandør: INTERSCIENCE
131025 132025 133025 132225 132525
INSC131025EA 1475 NOK
INSC131025 INSC132025 INSC133025 INSC132225 INSC132525
Blender bags, BagLight®
Poser Homogenisator-poser
These non-filter sample preparation bags are ideal for homogenising samples.
  • Robust/ultra resistant
  • Resistant to freezing
  • Compatible with any blender
  • Sterile and approved for food contact
BagLight® PolySilk®: Made of PolySilk® polyolefin complex, flexible and transparent. Available for different volume.
BagLight® Multilayer®: Made of Multilayer® reinforced complex, reinforced weldings.
BagLight® PolySilk® HD: Made of high density PolySilk®. They are rigid, matte and slightly opaque.