Colony Counter, Scan 50
Leverandør: INTERSCIENCE
435050
INSC435050EA 16400 NOK
INSC435050
Colony Counter, Scan 50
Kolonitellere
Scan 50 is a manual colony counter with improved ergonomics that help prevent musculoskeletal disorders.
  • Touch surface with adjustable sensitivity
  • Contrasting colony lighting: Dark field 2
  • Easy to clean and disinfect
  • Compatible with all types of media
  • Ergonomics redesigned
  • Nine different sounds and headphones jack to work without disturbing colleagues
  • Light signal for each counted colony
  • Optimal working posture, inclined reading surface with magnifying lens (optional), hand rest
  • Ambidextrous- duplicated count buttons on both side
It offers contrastingMore Product Information