High technology colony counter, Scan® 500
Leverandør: INTERSCIENCE
436000
INSC436000EA 0 NOK
INSC436000
High technology colony counter, Scan® 500
Kolonitellere
High technology colony counter with software that can be linked to a PC via a FireWire connection.
  • Minimum size of colonies is 0,1 mm
  • Detects up to seven colours on the same Petri dish
  • Measurement of the inhibition zone
Can count colonies on a Petri dish in less than one second for a fast, accurate, and traceable reading of the result.