Automatic colony counter, Scan® 300
Leverandør: INTERSCIENCE
436300
INSC436300EA 0 NOK
INSC436300
Automatic colony counter, Scan® 300
Kolonitellere
Automatic colony counter with a digital camera and high technology software.
  • Can be linked to a PC via a FireWire connection
  • Minimum size of colonies: 0,1 mm
  • Count all colonies in less than one second for a fast, accurate, and traceable reading of the result